Please login to your online account to display your discounted pricing

BDNF Polyclonal Antibody, Invitrogen™


BDNF Antibody, PA1-18375, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Western Blot.

Manufacturer: pierce biotechnology inc  PA118375

Catalog No. 01-670-627

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Western Blotting
Gene Symbols BDNF
Species Reactivity Human, Mouse, Rat
Storage Requirements Avoid Freeze/Thaw Cycles
Format Serum
Gene Accession No. P21237, P23363, P23560
Isotype IgG
Gene ID (Entrez) 12064, 24225, 627
Gene Alias Brain-derived neurotrophic factor, Abrineurin
Antigen BDNF
Quantity 100μL
Conjugate Unlabeled
Immunogen Synthetic peptide corresponding to residues H(129) S D P A R R G E L S V C D S I S E W V(148) of human BDNF protein.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit

BDNF Antibody, PA1-18375, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Western Blot.

plays a role in the development of hippocampal long term potentiation; involved in regulation of synaptic plasticity.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues H(129) S D P A R R G E L S V C D S I S E W V(148) of human BDNF protein.
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Brain-derived neurotrophic factor, Abrineurin
Uniprot Accession: P21237, P23363, P23560
Entrez GeneID: 12064, 24225, 627
Gene Symbol: BDNF
PA1-18375 detects BDNF from rat, human, and mouse samples. PA1-18375 has been successfully used in Western blot procedures. The PA1-18375 immunogen is a synthetic peptide corresponding to residues H(129) S D P A R R G E L S V C D S I S E W V(148) of human BDNF protein, conjugated to KLH. Reconstitute with 100 ul of distilled water.