Please login to your online account to display your discounted pricing

anti-BDNF Polyclonal, Thermo Scientific™

BDNF Polyclonal Ab PA1-18371 from Thermo Scientific™ specifically detects BDNF in Hu and Rt samples, and it is validated for ELISA, IHC and WB

$506.05 - $578.55

Specifications

Antigen BDNF
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-616

 
pierce biotechnology inc
PA118364
Unlabeled 100μL Each for $578.55

01-670-624

 
pierce biotechnology inc
PA118372
Unlabeled 500μg Each for $543.75

01-670-623

 
pierce biotechnology inc
PA118371
Unlabeled 100μL Each for $506.05
Description & Specifications

Specifications

Antigen BDNF
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Sheep
Immunogen Recombinant human BDNF.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

BDNF Polyclonal Ab PA1-18371 from Thermo Scientific™ specifically detects BDNF in Hu and Rt samples, and it is validated for ELISA, IHC and WB

BDNF belongs to the neurotrophin family and regulates the survival and differentiation of neurons during development. The alterations in BDNF expression induced by various kinds of brain insult including stress, ischemia, seizure activity and hypoglycemia, may contribute to some pathologies such as depression, epilepsy, Alzheimer's, and Parkinson's disease. BDNF promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. It is a major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability.
Host Species: Rabbit
Cross Reactivity: Human, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues H(131) S D P A R R G E L(140) of mouse BDNF.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Brain-derived neurotrophic factor, Abrineurin
Uniprot Accession: P23363, P23560
Gene Symbol: BDNF
Entrez GeneID: 24225, 627
PA1-18371 detects BDNF in Human and Rat samples. PA1-18371 has been successfully used in ELISA, Immunohistochemistry and Western Blot procedures. PA1-18371 The immunogen is Synthetic peptide corresponding to residues H(131) S D P A R R G E L(140) of mouse BDNF..