Please login to your online account to display your discounted pricing

anti-Bcl3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056375SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056375T

  • / Each
 

Description & Specifications

Specifications

Test Specificity In Jurkat, a 45 kDa band is observed. Cross reacts with Human. Predicted to cross react with Mouse and Rat.
Concentration 0.5mg/mL
Cross Reactivity Human
Formulation PBS containing 0.05% BSA
Gene ID (Entrez) 602
Gene Accession No. P20749
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Antigen Bcl3
Immunogen This antibody was made against synthetic peptides corresponding to amino acids 2-13 and 107-120 of human Bcl-3. It will cross-react with mouse and rat
Quantity 0.025mg
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Host Species Rabbit