Please login to your online account to display your discounted pricing

anti-Bcl3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056375

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056375

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Primary or Secondary Primary
Host Species Rabbit
Test Specificity In Jurkat, a 45 kDa band is observed. Cross reacts with Human. Predicted to cross react with Mouse and Rat.
Gene ID (Entrez) 602
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg
Gene Accession No. P20749
Regulatory Status RUO
Immunogen This antibody was made against synthetic peptides corresponding to amino acids 2-13 and 107-120 of human Bcl-3. It will cross-react with mouse and rat
Antigen Bcl3
Cross Reactivity Human
Applications Western Blotting
Format Purified