Please login to your online account to display your discounted pricing

anti-Bcl3, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was made against synthetic peptides corresponding to amino acids 2-13 and 107-120 of human Bcl-3. It will cross-react with mouse and rat
Antigen Bcl3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056375

 
novus biologicals canada
NB10056375
Unlabeled 0.1mg Each for $255.80

NB10056375T

 
novus biologicals canada
NB10056375SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was made against synthetic peptides corresponding to amino acids 2-13 and 107-120 of human Bcl-3. It will cross-react with mouse and rat
Antigen Bcl3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity In Jurkat, a 45 kDa band is observed. Cross reacts with Human. Predicted to cross react with Mouse and Rat.
Gene Accession No. P20749
Gene ID (Entrez) 602
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL