Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056103SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056103T

  • / Each
 

Description & Specifications

Specifications

Test Specificity Bcl-XL + S. Dog, Gerbil, Human, Mouse, Rat. Not yet tested in other species.
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 598
Primary or Secondary Primary
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Antigen Bcl-xL
Conjugate Unlabeled
Host Species Rabbit
Immunogen A recombinant human Bcl-XL protein without the N-terminal transmembrane portion was used as immunogen.
Format Antiserum