Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056103

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056103

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 598
Test Specificity Bcl-XL + S. Dog, Gerbil, Human, Mouse, Rat. Not yet tested in other species.
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Immunogen A recombinant human Bcl-XL protein without the N-terminal transmembrane portion was used as immunogen.
Quantity 0.05mL
Format Antiserum
Primary or Secondary Primary
Host Species Rabbit