Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11083982SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11083982T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 598
Concentration 1mg/mL
Format Affinity Purified
Gene Accession No. Q07817
Formulation Tris-citrate/phosphate, pH 7-8
Monoclonal or Polyclonal Polyclonal
Test Specificity There are three known isoforms of Bcl-X; Bcl-X(S), Bcl-X(beta), and Bcl-X(L). The immunogen used to create this has 100% identity to all three isoforms. Human, feline, canine and sheep. Immunogen sequence has 93% homology to porcine, 87% to mouse and 81% to rat.
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Sheep
Quantity 0.025mL
Regulatory Status RUO
Host Species Rabbit
Conjugate Unlabeled
Antigen Bcl-xL
Primary or Secondary Primary
Immunogen Synthetic peptide made to an N-terminal portion of human BCL2L1 (within residues 30-70). [Swiss-Prot No. Q07817]
Applications Western Blotting