Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056312SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056312T

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-xL
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS
Gene Accession No. NP_612815
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 3-14 that is common in human and mouse Bcl-XL and Bcl-XS protein was used as immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 598
Test Specificity Cross reacts with Human. Predicted to cross react with Mouse.
Target Species Human