Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056312

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056312

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene ID (Entrez) 598
Test Specificity Cross reacts with Human. Predicted to cross react with Mouse.
Concentration 0.5mg/mL
Conjugate Unlabeled
Gene Accession No. NP_612815
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Antigen Bcl-xL
Quantity 0.1mg
Regulatory Status RUO
Immunogen A synthetic peptide corresponding to amino acids 3-14 that is common in human and mouse Bcl-XL and Bcl-XS protein was used as immunogen.
Cross Reactivity Human
Format Purified
Primary or Secondary Primary
Applications Western Blotting