Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 3-14 that is common in human and mouse Bcl-XL and Bcl-XS protein was used as immunogen.
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056312T

 
novus biologicals canada
NB10056312SS
Unlabeled 0.025mg Each for $139.00

NB10056312

 
novus biologicals canada
NB10056312
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 3-14 that is common in human and mouse Bcl-XL and Bcl-XS protein was used as immunogen.
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Cross reacts with Human. Predicted to cross react with Mouse.
Gene Accession No. NP_612815
Gene ID (Entrez) 598
Formulation PBS
Concentration 0.5mg/mL