Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 61-79 (DSPAVNGATAHSSSLDARE) of human Bcl-XL was used as immunogen; GenBank no. CAA80661.1. The immunogen sequence is 94% conserved in Bcl-XS.
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056105

 
novus biologicals canada
NB10056105
Unlabeled 0.05mL Each for $222.95

NB10056105T

 
novus biologicals canada
NB10056105SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 61-79 (DSPAVNGATAHSSSLDARE) of human Bcl-XL was used as immunogen; GenBank no. CAA80661.1. The immunogen sequence is 94% conserved in Bcl-XS.
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Bcl-XL + S. Gerbil, Human, Mouse, Rat Not yet tested in other species.
Gene ID (Entrez) 598
Formulation PBS