Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 46-66 (ETPSAINGNPSWHLADSPAVN) of human Bcl-XL was used as immunogen; GenBank no. CAA80661.1. The immunogen sequence is 100% conserved in Bcl-XS. The immunogen sequence is 100% in multiple species including mouse, rat, dog, cow, gerbil, and sheep.
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056104

 
novus biologicals canada
NB10056104
Unlabeled 0.05mL Each for $222.95

NB10056104T

 
novus biologicals canada
NB10056104SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 46-66 (ETPSAINGNPSWHLADSPAVN) of human Bcl-XL was used as immunogen; GenBank no. CAA80661.1. The immunogen sequence is 100% conserved in Bcl-XS. The immunogen sequence is 100% in multiple species including mouse, rat, dog, cow, gerbil, and sheep.
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Bcl-XL + S. The immunogen sequence is 100% in multiple species including mouse, rat, dog, cow, gerbil, and sheep.
Gene ID (Entrez) 598