Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A recombinant human Bcl-XL protein without the N-terminal transmembrane portion was used as immunogen.
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056103T

 
novus biologicals canada
NB10056103SS
Unlabeled 0.025mL Each for $144.63

NB10056103

 
novus biologicals canada
NB10056103
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A recombinant human Bcl-XL protein without the N-terminal transmembrane portion was used as immunogen.
Antigen Bcl-xL
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Bcl-XL + S. Dog, Gerbil, Human, Mouse, Rat. Not yet tested in other species.
Gene ID (Entrez) 598