Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056105SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056105T

  • / Each
 

Description & Specifications

Specifications

Test Specificity Bcl-XL + S. Gerbil, Human, Mouse, Rat Not yet tested in other species.
Immunogen A synthetic peptide corresponding to amino acids 61-79 (DSPAVNGATAHSSSLDARE) of human Bcl-XL was used as immunogen; GenBank no. CAA80661.1. The immunogen sequence is 94% conserved in Bcl-XS.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Conjugate Unlabeled
Gene ID (Entrez) 598
Formulation PBS
Primary or Secondary Primary
Regulatory Status RUO
Host Species Rabbit
Antigen Bcl-xL
Format Antiserum
Monoclonal or Polyclonal Polyclonal
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mL