Please login to your online account to display your discounted pricing

anti-Bcl-xL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056105

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056105

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Test Specificity Bcl-XL + S. Gerbil, Human, Mouse, Rat Not yet tested in other species.
Formulation PBS
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene ID (Entrez) 598
Immunogen A synthetic peptide corresponding to amino acids 61-79 (DSPAVNGATAHSSSLDARE) of human Bcl-XL was used as immunogen; GenBank no. CAA80661.1. The immunogen sequence is 94% conserved in Bcl-XS.
Antigen Bcl-xL
Primary or Secondary Primary
Conjugate Unlabeled
Quantity 0.05mL
Format Antiserum
Regulatory Status RUO