Please login to your online account to display your discounted pricing

anti-Bcl rambo, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056663

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056663

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_056182
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23786
Applications Western Blotting
Conjugate Unlabeled
Formulation PBS containing 0.2% gelatin
Format Antiserum
Test Specificity In HeLa, a 56 kDa band is observed.
Antigen Bcl rambo
Regulatory Status RUO
Cross Reactivity Human
Host Species Rabbit
Primary or Secondary Primary
Immunogen This antibody was developed against a synthetic peptide corresponding to a portion of human Bcl-rambo.
Quantity 0.2mL