Please login to your online account to display your discounted pricing

anti-Bcl rambo, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to a portion of human Bcl-rambo.
Antigen Bcl rambo
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056663T

 
novus biologicals canada
NB10056663SS
Unlabeled 0.05mL Each for $139.00

NB10056663

 
novus biologicals canada
NB10056663
Unlabeled 0.2mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to a portion of human Bcl-rambo.
Antigen Bcl rambo
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In HeLa, a 56 kDa band is observed.
Gene Accession No. NP_056182
Gene ID (Entrez) 23786
Formulation PBS containing 0.2% gelatin