Please login to your online account to display your discounted pricing

anti-Bcl rambo, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056663SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056663T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Test Specificity In HeLa, a 56 kDa band is observed.
Gene Accession No. NP_056182
Gene ID (Entrez) 23786
Conjugate Unlabeled
Immunogen This antibody was developed against a synthetic peptide corresponding to a portion of human Bcl-rambo.
Regulatory Status RUO
Antigen Bcl rambo
Cross Reactivity Human
Applications Western Blotting
Format Antiserum
Primary or Secondary Primary
Quantity 0.05mL