Please login to your online account to display your discounted pricing

anti-Bcl-2 related protein A1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056070SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056070T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 597
Test Specificity A1 . Cross reacts with Gerbil, Mouse and Rat.
Primary or Secondary Primary
Format Antiserum
Formulation PBS
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mL
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Bcl-2 related protein A1
Immunogen A synthetic peptide corresponding to amino acids (aa) 34-51 (CRVLQRVAFSVQKEVEKN) of mouse A1 was used as immunogen; GenBank no. AAH28762.1 Mouse A1 is a 172 aa acid protein.
Conjugate Unlabeled
Host Species Rabbit