Please login to your online account to display your discounted pricing

anti-Bcl-2 related protein A1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056070

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056070

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene ID (Entrez) 597
Conjugate Unlabeled
Quantity 0.05mL
Cross Reactivity Mouse
Cross Reactivity Rat
Test Specificity A1 . Cross reacts with Gerbil, Mouse and Rat.
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Antigen Bcl-2 related protein A1
Format Antiserum
Immunogen A synthetic peptide corresponding to amino acids (aa) 34-51 (CRVLQRVAFSVQKEVEKN) of mouse A1 was used as immunogen; GenBank no. AAH28762.1 Mouse A1 is a 172 aa acid protein.
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary