Please login to your online account to display your discounted pricing

anti-Bcl-2 related protein A1, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids (aa) 34-51 (CRVLQRVAFSVQKEVEKN) of mouse A1 was used as immunogen; GenBank no. AAH28762.1 Mouse A1 is a 172 aa acid protein.
Antigen Bcl-2 related protein A1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056070

 
novus biologicals canada
NB10056070
Unlabeled 0.05mL Each for $222.95

NB10056070T

 
novus biologicals canada
NB10056070SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids (aa) 34-51 (CRVLQRVAFSVQKEVEKN) of mouse A1 was used as immunogen; GenBank no. AAH28762.1 Mouse A1 is a 172 aa acid protein.
Antigen Bcl-2 related protein A1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity A1 . Cross reacts with Gerbil, Mouse and Rat.
Gene ID (Entrez) 597
Formulation PBS