Please login to your online account to display your discounted pricing

anti-Bcl-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056099SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056099T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Quantity 0.025mL
Test Specificity Bcl-2 . Reacts with Feline, Canine and Human. 92% sequence identity with bovine and 84% with mouse and rat proteins.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Regulatory Status RUO
Gene ID (Entrez) 596
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Immunogen 92% conserved in cow Bcl-2 and 84% conserved in mouse and rat Bcl-2.
Host Species Rabbit
Antigen Bcl-2
Format Antiserum