Please login to your online account to display your discounted pricing

anti-Bcl-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056099

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056099

  • / Each
 

Description & Specifications

Specifications

Quantity 0.05mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Gene ID (Entrez) 596
Test Specificity Bcl-2 . Reacts with Feline, Canine and Human. 92% sequence identity with bovine and 84% with mouse and rat proteins.
Immunogen 92% conserved in cow Bcl-2 and 84% conserved in mouse and rat Bcl-2.
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Format Antiserum
Antigen Bcl-2
Host Species Rabbit