Please login to your online account to display your discounted pricing

anti-Bcl-2, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen 92% conserved in cow Bcl-2 and 84% conserved in mouse and rat Bcl-2.
Antigen Bcl-2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056099

 
novus biologicals canada
NB10056099
Unlabeled 0.05mL Each for $222.95

NB10056099T

 
novus biologicals canada
NB10056099SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 92% conserved in cow Bcl-2 and 84% conserved in mouse and rat Bcl-2.
Antigen Bcl-2
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Bcl-2 . Reacts with Feline, Canine and Human. 92% sequence identity with bovine and 84% with mouse and rat proteins.
Gene ID (Entrez) 596