Please login to your online account to display your discounted pricing

anti-BAK, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056092SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056092T

  • / Each
 

Description & Specifications

Specifications

Antigen BAK
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation PBS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 16-36 (EPALPSASEEQVAQDTEEVFR) of human Bak was used as immunogen
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 578
Test Specificity BAK .
Target Species Human