Please login to your online account to display your discounted pricing

anti-BAK, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056092

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056092

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Primary or Secondary Primary
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.05mL
Gene ID (Entrez) 578
Test Specificity BAK .
Regulatory Status RUO
Immunogen A synthetic peptide corresponding to amino acids 16-36 (EPALPSASEEQVAQDTEEVFR) of human Bak was used as immunogen
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Format Antiserum
Host Species Rabbit
Conjugate Unlabeled
Antigen BAK