Please login to your online account to display your discounted pricing

anti-BAK, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 16-36 (EPALPSASEEQVAQDTEEVFR) of human Bak was used as immunogen
Antigen BAK
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056092

 
novus biologicals canada
NB10056092
Unlabeled 0.05mL Each for $222.95

NB10056092T

 
novus biologicals canada
NB10056092SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 16-36 (EPALPSASEEQVAQDTEEVFR) of human Bak was used as immunogen
Antigen BAK
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity BAK .
Gene ID (Entrez) 578
Formulation PBS