Please login to your online account to display your discounted pricing

anti-BAG5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056091SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056091T

  • / Each
 

Description & Specifications

Specifications

Antigen BAG5
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antisera
Formulation PBS
Host Species Rabbit
Immunogen 95% conserved with mouse BAG-5
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9529
Test Specificity BAG5 .
Target Species Bovine
Target Species Canine
Target Species Human
Target Species Murine
Target Species Rat