Please login to your online account to display your discounted pricing

anti-BAG5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056091

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056091

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Immunogen 95% conserved with mouse BAG-5
Gene ID (Entrez) 9529
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.05mL
Test Specificity BAG5 .
Antigen BAG5
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Antiserum
Conjugate Unlabeled
Host Species Rabbit