Please login to your online account to display your discounted pricing

anti-BAG5, Polyclonal, Novus Biologicals™

$144.63 - $252.00

Specifications

Host Species Rabbit
Immunogen 95% conserved with mouse BAG-5
Antigen BAG5
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056091T

 
novus biologicals canada
NB10056091SS
Unlabeled 0.025mL Each for $144.63

NB10056091

 
novus biologicals canada
NB10056091
Unlabeled 0.05mL Each for $252.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 95% conserved with mouse BAG-5
Antigen BAG5
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity BAG5 .
Gene ID (Entrez) 9529
Formulation PBS