Please login to your online account to display your discounted pricing

anti-Bag-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056085

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056085

  • / Each
 

Description & Specifications

Specifications

Antigen Bag-1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antisera
Formulation PBS
Host Species Rabbit
Immunogen A synthethic BAG-1 peptide was used as immunogen
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 573
Test Specificity BAG1 .
Target Species Human
Target Species Murine
Target Species Rat