Please login to your online account to display your discounted pricing

anti-Bag-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056084SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056084T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 573
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation PBS
Test Specificity BAG1 .
Conjugate Unlabeled
Quantity 0.025mL
Antigen Bag-1
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Full-length recombinant mouse BAG-1 protein was used as immunogen.
Host Species Rabbit
Format Antiserum