Please login to your online account to display your discounted pricing

anti-Bag-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056083SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056083T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity BAG1 .
Formulation PBS
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 573
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Full-length recombinant mouse BAG-1 protein was used as immunogen.
Host Species Rabbit
Quantity 0.025mL
Antigen Bag-1
Format Antiserum