Please login to your online account to display your discounted pricing

anti-Bag-1, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthethic BAG-1 peptide was used as immunogen
Antigen Bag-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056085T

 
novus biologicals canada
NB10056085SS
Unlabeled 0.025mL Each for $144.63

NB10056085

 
novus biologicals canada
NB10056085
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthethic BAG-1 peptide was used as immunogen
Antigen Bag-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity BAG1 .
Gene ID (Entrez) 573
Formulation PBS