Please login to your online account to display your discounted pricing

anti-Bag-1, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen Full-length recombinant mouse BAG-1 protein was used as immunogen.
Antigen Bag-1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056084T

 
novus biologicals canada
NB10056084SS
Unlabeled 0.025mL Each for $144.63

NB10056084

 
novus biologicals canada
NB10056084
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full-length recombinant mouse BAG-1 protein was used as immunogen.
Antigen Bag-1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity BAG1 .
Gene ID (Entrez) 573
Formulation PBS