Please login to your online account to display your discounted pricing

anti-Bad, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corrsponding to amino acids 87-103 (GEEPSPFRGRSRSAPPN) of human Bad was used as immunogen; GenBank no. NP_004313.1. Amino acids 88-103 of the peptide immunogen are 93% conserved in mouse and rat Bad.
Antigen Bad
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056078T

 
novus biologicals canada
NB10056078SS
Unlabeled 0.025mL Each for $144.63

NB10056078

 
novus biologicals canada
NB10056078
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corrsponding to amino acids 87-103 (GEEPSPFRGRSRSAPPN) of human Bad was used as immunogen; GenBank no. NP_004313.1. Amino acids 88-103 of the peptide immunogen are 93% conserved in mouse and rat Bad.
Antigen Bad
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Bad . Reacts with Human. 93% sequence identity with mouse and rat proteins.
Gene ID (Entrez) 572
Formulation PBS