Please login to your online account to display your discounted pricing

anti-B220/CD45R, Clone: RA3-6B2, R&D Systems

$139.08 - $432.57

Specifications

Antigen B220/CD45R
Clone RA3-6B2
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB1217

 
r & d systems
RDSMAB1217
Unlabeled 500μg Each for $432.57

MLDP7

 
r & d systems
RDSMLDP7
Unlabeled 1 Vial Each for $139.08
Description & Specifications

Specifications

Antigen B220/CD45R
Clone RA3-6B2
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Abelson mouse leukemia virus-induced pre-B tumor cells.
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Cell Depletion