Please login to your online account to display your discounted pricing

anti-B220/CD45R, Clone: RA3-6B2, R&D Systems


Manufacturer: r & d systems  RDSMLDP7

 View more versions of this product

Catalog No. MLDP7

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Immunogen Abelson mouse leukemia virus-induced pre-B tumor cells.
Host Species Rat
Clone RA3-6B2
Quantity 1 Vial
Format Affinity Purified
Conjugate Unlabeled
Cross Reactivity Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Isotype IgG2a
Antigen B220/CD45R