Please login to your online account to display your discounted pricing

anti-Axl, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224497SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224497SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 558
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, zebrafish, xenopus and chicken.
Gene Accession No. P30530
Monoclonal or Polyclonal Polyclonal
Immunogen A portion of amino acids 450-500 of human AXL/UFON was used as the immunogen.
Formulation 100μg in 200μL PBS containing 0.05% BSA
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Concentration 0.5mg/mL
Regulatory Status RUO
Applications Western Blotting
Format Affinity Purified
Host Species Rabbit
Quantity 0.025mg
Antigen Axl
Conjugate Unlabeled
Primary or Secondary Primary