Please login to your online account to display your discounted pricing

anti-Axl, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224497

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224497

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. P30530
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 558
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, zebrafish, xenopus and chicken.
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Immunogen A portion of amino acids 450-500 of human AXL/UFON was used as the immunogen.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Axl
Applications Western Blotting
Quantity 0.1mg
Host Species Rabbit
Regulatory Status RUO
Primary or Secondary Primary