Please login to your online account to display your discounted pricing

anti-Axl, Polyclonal, Novus Biologicals™

$139.00 - $139.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 450-500 of human AXL/UFON was used as the immunogen.
Antigen Axl
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224497SS

 
novus biologicals canada
NBP224497SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 450-500 of human AXL/UFON was used as the immunogen.
Antigen Axl
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, zebrafish, xenopus and chicken.
Gene Accession No. P30530
Gene ID (Entrez) 558
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL