Please login to your online account to display your discounted pricing

anti-Avian Flu HA protein, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Amino acids 128-143 of avian flu HA protein were used as the immunogen.
Antigen Avian Flu HA protein
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056608T

 
novus biologicals canada
NB10056608SS
Unlabeled 0.025mg Each for $139.00

NB10056608

 
novus biologicals canada
NB10056608
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 128-143 of avian flu HA protein were used as the immunogen.
Antigen Avian Flu HA protein
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity This will not cross with the H5N1 strain.
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL