Please login to your online account to display your discounted pricing

anti-Autotaxin Polyclonal, Thermo Scientific™

Autotaxin Polyclonal antibody PA1-12732 from Thermo Scientific™ specifically detects Autotaxin in Human samples, and it is validated for ELISA and Western Blot

$797.50 - $797.50

Specifications

Antigen Autotaxin
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to autotaxin.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-858

 
pierce biotechnology inc
PA112732
Unlabeled 100μg Each for $797.50
Description & Specifications

Specifications

Antigen Autotaxin
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to autotaxin.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Autotaxin Polyclonal antibody PA1-12732 from Thermo Scientific™ specifically detects Autotaxin in Human samples, and it is validated for ELISA and Western Blot

Autotaxin (ATX), a tumor cell motility- stimulating protein is an ecto/exo-enzyme with significant homology to the plasma cell membrane differentiation antigen PC-1. ATX is a 99kDa core protein and may run as a 125-kDa glycoprotein when isolated from the human melanoma cell line (A2058). ATX, like PC-1 can hydrolyze the type I phosphodiesterase substrate p-nitrophenyl thymidine-5'-monophosphate. Autotaxin has a novel motilityregulating function for this class of ecto/exo-enzymes.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic peptide corresponding to autotaxin.
Formulation: 100μg purified Antibody (1mg/mL) in PBS, with 0.08% sodium azide;
Synonyms: ATX, ATX-X, AUTOTAXIN, FLJ26803, LysoPLD, NPP2, PD-IALPHA, PDNP2
Uniprot Accession: Q13822
Gene Symbol: ENPP2
Entrez GeneID: 5168
PA1-12732 detects Autotaxin in Human samples. PA1-12732 has been successfully used in ELISA and Western Blot procedures. PA1-12732 The immunogen is Synthetic peptide corresponding to autotaxin..