Please login to your online account to display your discounted pricing

anti-Autoimmune Regulator/AIRE, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224624SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224624SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. O43918
Immunogen A portion of amino acids 20-70 of human AIRE was used as the immunogen.
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee.
Antigen Autoimmune Regulator/AIRE
Gene ID (Entrez) 326
Concentration 0.5mg/mL
Applications Western Blotting
Format Purified
Quantity 0.025mg
Primary or Secondary Primary
Host Species Rabbit
Regulatory Status RUO