missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Autoimmune Regulator/AIRE, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 20-70 of human AIRE was used as the immunogen.
Antigen Autoimmune Regulator/AIRE
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224624SS Novus Biologicals Canada
NBP224624SS
0.025mg Each for $109.00
NBP224624 Novus Biologicals Canada
NBP224624
0.1mg Each for $257.41
Specifications Description & Specifications

Specifications

Rabbit
Autoimmune Regulator/AIRE
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee.
326
0.5mg/mL
A portion of amino acids 20-70 of human AIRE was used as the immunogen.
Polyclonal
Primary
Western Blot
O43918
100μg in 200μL PBS containing 0.05% BSA