Please login to your online account to display your discounted pricing

anti-Aurora A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP151843

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP151843

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Concentration 0.56mg/mL
Formulation PBS, 0.1% BSA, 50% glycerol
Gene Accession No. O14965
Conjugate Unlabeled
Gene ID (Entrez) 6790
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Aurora A
Host Species Rabbit
Quantity 0.1mL
Immunogen A genomic peptide made to an N-terminal region of the human Aurora A protein (within residues 50-200). [Swiss-Prot O14965]
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified