Please login to your online account to display your discounted pricing

anti-Aurora A, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human Aurora A protein (within residues 50-200). [Swiss-Prot O14965]
Antigen Aurora A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP151843

 
novus biologicals canada
NBP151843
Unlabeled 0.1mL Each for $329.00

NBP151843SS

 
novus biologicals canada
NBP151843SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human Aurora A protein (within residues 50-200). [Swiss-Prot O14965]
Antigen Aurora A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Gene Accession No. O14965
Gene ID (Entrez) 6790
Formulation PBS, 0.1% BSA, 50% glycerol
Concentration 0.56mg/mL