Please login to your online account to display your discounted pricing

anti-AU5 Epitope Tag, Polyclonal, Novus Biologicals™

Manufacturer: novus biologicals canada  NB600-458-1mg

 View all options for this product


For Research Use Only

Catalog No. NB6004581MG


Description & Specifications


Antigen AU5 Epitope Tag
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Non-species specific
Format Affinity Purified
Formulation phosphate Buffered Saline (PBS)
Host Species Rabbit
Immunogen Rabbits were immunized with AU5 (TDFYLK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Quantity 1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal