Please login to your online account to display your discounted pricing

anti-AU5 Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6004611MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6004611MG

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Concentration 1.0mg/mL
Immunogen Goats were immunized with AU5 (TDFYLK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Formulation phosphate Buffered Saline (PBS)
Regulatory Status RUO
Host Species Goat
Quantity 1mg
Antigen AU5 Epitope Tag
Cross Reactivity Non-species specific
Format Affinity Purified